Assay Details
| Gene Symbol: Tsnax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022262.3 | NP_071598.1 | 3 - 4 | 313 | 98 | |
| GenBank mRNA | AF262357.1 | - | 3 - 4 | 384 | 98 | |
| BC081715.1 | - | 3 - 4 | 313 | 7130295 | 98 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tsnax
Genomic Map