Assay Details
| Gene Symbol: Tomm20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152935.1 | NP_690918.1 | 2 - 3 | 273 | 137 | |
| GenBank mRNA | BC078715.1 | - | 2 - 3 | 249 | 7127457 | 137 |
| D63411.1 | - | 2 - 3 | 262 | 137 | ||
| FQ224957.1 | - | 2 - 3 | 290 | 137 | ||
| U21871.1 | - | 2 - 3 | 273 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tomm20
Genomic Map