Assay Details
| Gene Symbol: Slc35f3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772680.1 | XP_008770902.1 | 1 - 2 | 344 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35f3| Gene Symbol: Slc35f3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772680.1 | XP_008770902.1 | 1 - 2 | 344 | 79 | |
Set Membership: |
|