Assay Details
| Gene Symbol: Rab4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013019.2 | NP_037151.2 | 2 - 3 | 238 | 70 | |
| XM_008772599.1 | XP_008770821.1 | 2 - 3 | 1077 | 70 | ||
| XM_008772600.1 | XP_008770822.1 | 2 - 3 | 289 | 70 | ||
| GenBank mRNA | BC062016.1 | - | 2 - 3 | 238 | 5621494 | 70 |
| X06890.1 | - | 2 - 3 | 170 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008772600.1 transcripts.
Set Membership: |
|

Rab4a
Genomic Map