Assay Details
| Gene Symbol: Gabarapl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022706.2 | NP_073197.1 | 1 - 2 | 100 | 71 | |
| GenBank mRNA | AB003515.1 | - | 1 - 2 | 138 | 71 | |
| BC058145.1 | - | 1 - 2 | 103 | 6921109 | 71 | |
| BC088139.1 | - | 1 - 2 | 100 | 7323806 | 71 | |
| FQ216852.1 | - | 1 - 2 | 114 | 71 | ||
| FQ226397.1 | - | 1 - 2 | 124 | 71 | ||
| FQ231643.1 | - | 1 - 2 | 93 | 71 | ||
| FQ233551.1 | - | 1 - 2 | 135 | 71 | ||
| FQ234861.1 | - | 1 - 2 | 93 | 71 | ||
| Gene Symbol: LOC100359937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729947.4 | XP_002729993.3 | 1 | 89 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB003515.1, BC058145.1, BC088139.1, NM_022706.2, XM_002729947.4 transcripts.
Set Membership: |
|

Gabarapl2
Genomic Map