Assay Details
| Gene Symbol: Gabarapl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022706.2 | NP_073197.1 | 2 - 3 | 160 | 122 | |
| GenBank mRNA | AB003515.1 | - | 2 - 3 | 198 | 122 | |
| BC058145.1 | - | 2 - 3 | 163 | 6921109 | 122 | |
| BC088139.1 | - | 2 - 3 | 160 | 7323806 | 122 | |
| FQ216852.1 | - | 2 - 3 | 174 | 122 | ||
| FQ226397.1 | - | 2 - 3 | 184 | 122 | ||
| FQ231643.1 | - | 2 - 3 | 153 | 122 | ||
| FQ233551.1 | - | 2 - 3 | 195 | 122 | ||
| FQ234861.1 | - | 2 - 3 | 153 | 122 | ||
| Gene Symbol: LOC100359937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729947.4 | XP_002729993.3 | 1 | 149 | 122 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gabarapl2
Genomic Map