Assay Details
| Gene Symbol: Wdr83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047847.1 | NP_001041312.1 | 4 - 5 | 443 | 89 | |
| GenBank mRNA | AY940050.1 | - | 4 - 5 | 443 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr83
Genomic Map