Assay Details
| Gene Symbol: Efna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053903.1 | NP_446355.1 | 2 - 3 | 459 | 104 | |
| XM_006245593.2 | XP_006245655.1 | 2 - 3 | 652 | 104 | ||
| GenBank mRNA | U69279.1 | - | 2 - 3 | 459 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Efna5
Genomic Map