Assay Details
| Gene Symbol: Farp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108233.1 | NP_001101703.1 | 22 - 23 | 2610 | 69 | |
| XM_006245529.2 | XP_006245591.1 | 23 - 24 | 2638 | 69 | ||
| XM_006245530.2 | XP_006245592.1 | 24 - 25 | 2714 | 69 | ||
| XM_006245532.2 | XP_006245594.1 | 22 - 23 | 2617 | 69 | ||
| XM_008767375.1 | XP_008765597.1 | 23 - 24 | 2571 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Farp2
Genomic Map