Assay Details
| Gene Symbol: Mterfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037209.1 | NP_001032286.1 | 2 - 3 | 522 | 96 | |
| GenBank mRNA | BC098676.1 | - | 2 - 3 | 522 | 7372734 | 96 |
More Information
Updated Mapping Information
Set Membership: |
|

Mterfd2
Genomic Map