Assay Details
| Gene Symbol: Rab31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145094.2 | NP_659562.2 | 1 - 2 | 131 | 87 | |
| GenBank mRNA | AF254800.1 | - | 1 - 2 | 80 | 87 | |
| BC072698.1 | - | 1 - 2 | 131 | 7105732 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab31
Genomic Map