Assay Details
| Gene Symbol: U2af1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256228.2 | XP_006256290.1 | 3 - 4 | 261 | 78 | |
| GenBank mRNA | FQ229339.1 | - | 3 - 4 | 238 | 78 | |
More Information
Set Membership: |
|

U2af1
Genomic Map