Assay Details
| Gene Symbol: RT1-Da | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008847.2 | NP_001008847.1 | 1 - 2 | 136 | 101 | |
| GenBank mRNA | BC098677.1 | - | 1 - 2 | 102 | 7373701 | 101 |
| FM048166.1 | - | 1 - 2 | 136 | 101 | ||
| FQ218574.1 | - | 1 - 2 | 127 | 101 | ||
| FQ220763.1 | - | 1 - 2 | 124 | 101 | ||
| FQ220966.1 | - | 1 - 2 | 129 | 101 | ||
| FQ226848.1 | - | 1 - 2 | 127 | 101 | ||
| FQ227693.1 | - | 1 - 2 | 127 | 101 | ||
| FQ227948.1 | - | 1 - 2 | 127 | 101 | ||
| FQ230207.1 | - | 1 - 2 | 127 | 101 | ||
| FQ232717.1 | - | 1 - 2 | 124 | 101 | ||
| FQ233042.1 | - | 1 - 2 | 127 | 101 | ||
| FQ233454.1 | - | 1 - 2 | 119 | 101 | ||
| FQ233943.1 | - | 1 - 2 | 127 | 101 | ||
| FQ234684.1 | - | 1 - 2 | 129 | 101 | ||
| FQ235025.1 | - | 1 - 2 | 119 | 101 | ||
| KC222932.1 | - | 1 - 2 | 81 | 101 | ||
| KC222933.1 | - | 1 - 2 | 81 | 101 | ||
| KC222934.1 | - | 1 - 2 | 81 | 101 | ||
| KC222935.1 | - | 1 - 2 | 81 | 101 | ||
| KC222936.1 | - | 1 - 2 | 81 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RT1-Da
Genomic Map