Assay Details
| Gene Symbol: Ubash3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107620.1 | NP_001101090.1 | 9 - 10 | 1638 | 81 | |
| XM_006256215.2 | XP_006256277.1 | 10 - 11 | 1762 | 81 | ||
| XM_008772818.1 | XP_008771040.1 | 10 - 11 | 1378 | 81 | ||
| XM_008772819.1 | XP_008771041.1 | 10 - 11 | 1373 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ubash3a
Genomic Map