Assay Details
| Gene Symbol: Cstb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012838.2 | NP_036970.1 | 1 - 2 | 167 | 145 | |
| GenBank mRNA | BC084725.1 | - | 1 - 2 | 155 | 7318993 | 145 |
| CF976177.1 | - | 1 - 2 | 168 | 145 | ||
| FQ221069.1 | - | 1 - 2 | 127 | 145 | ||
| FQ222613.1 | - | 1 - 2 | 122 | 145 | ||
| X54737.1 | - | 1 - 2 | 159 | 145 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cstb
Genomic Map