Assay Details
| Gene Symbol: Icoslg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256260.2 | XP_006256322.1 | 3 - 4 | 642 | 102 | |
| XM_006256261.2 | XP_006256323.1 | 3 - 4 | 639 | 102 | ||
| XM_006256262.2 | XP_006256324.1 | 3 - 4 | 641 | 102 | ||
| XM_006256264.2 | XP_006256326.1 | 3 - 4 | 640 | 102 | ||
More Information
Set Membership: |
|

Icoslg
Genomic Map