Assay Details
| Gene Symbol: Psmb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012708.2 | NP_036840.2 | 3 - 4 | 306 | 138 | |
| GenBank mRNA | BC091161.1 | - | 3 - 4 | 294 | 7366097 | 138 |
| BP500440.1 | - | 3 - 4 | 311 | 138 | ||
| D10757.1 | - | 3 - 4 | 273 | 138 | ||
| FQ228039.1 | - | 3 - 4 | 280 | 138 | ||
| KC222887.1 | - | 3 - 4 | 255 | 138 | ||
| KC222888.1 | - | 3 - 4 | 255 | 138 | ||
| KC222889.1 | - | 3 - 4 | 255 | 138 | ||
| KC222890.1 | - | 3 - 4 | 255 | 138 | ||
| KC222891.1 | - | 3 - 4 | 255 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmb9
Genomic Map