Assay Details
| Gene Symbol: Pttg1ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013238.1 | NP_001013256.1 | 2 - 3 | 223 | 76 | |
| XM_006256253.2 | XP_006256315.1 | 3 - 4 | 413 | 76 | ||
| GenBank mRNA | BC061813.1 | - | 2 - 3 | 223 | 5598663 | 76 |
| FQ218172.1 | - | 2 - 3 | 235 | 76 | ||
| FQ228848.1 | - | 2 - 3 | 261 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pttg1ip
Genomic Map