Assay Details
| Gene Symbol: Gstt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109675.1 | NP_001103145.1 | 1 - 2 | 218 | 150 | |
| GenBank mRNA | BC097423.1 | - | 1 - 2 | 202 | 7455039 | 150 |
More Information
Updated Mapping Information
Set Membership: |
|

Gstt4
Genomic Map