Assay Details
| Gene Symbol: Hsd17b8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212529.1 | NP_997694.1 | 5 - 6 | 576 | 122 | |
| XM_006255964.2 | XP_006256026.1 | 4 - 5 | 654 | 122 | ||
| XM_006255965.2 | XP_006256027.1 | 5 - 6 | 691 | 122 | ||
| XM_006255966.2 | XP_006256028.1 | 4 - 5 | 654 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b8
Genomic Map