Assay Details
| Gene Symbol: Rab36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109589.2 | NP_001103059.1 | 4 - 5 | 287 | 70 | |
| XM_006256342.2 | XP_006256404.1 | 4 - 5 | 717 | 70 | ||
| XM_008772925.1 | XP_008771147.1 | 4 - 5 | 722 | 70 | ||
| XM_008772926.1 | XP_008771148.1 | 4 - 5 | 723 | 70 | ||
| GenBank mRNA | BC167029.1 | - | 4 - 5 | 287 | 8368465 | 70 |
| FQ213589.1 | - | 3 - 4 | 213 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab36
Genomic Map