Assay Details
| Gene Symbol: Rps18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213557.1 | NP_998722.1 | 1 - 2 | 46 | 62 | |
| GenBank mRNA | FQ209845.1 | - | 1 - 2 | 44 | 62 | |
| FQ210792.1 | - | 1 - 2 | 44 | 62 | ||
| FQ220696.1 | - | 1 - 2 | 44 | 62 | ||
| FQ221730.1 | - | 1 - 2 | 62 | |||
| FQ221918.1 | - | 1 - 2 | 44 | 62 | ||
| FQ222652.1 | - | 1 - 2 | 44 | 62 | ||
| FQ222675.1 | - | 1 - 2 | 42 | 62 | ||
| FQ223127.1 | - | 1 - 2 | 44 | 62 | ||
| FQ223169.1 | - | 1 - 2 | 45 | 62 | ||
| FQ223198.1 | - | 1 - 2 | 44 | 62 | ||
| FQ223436.1 | - | 1 - 2 | 44 | 62 | ||
| FQ223481.1 | - | 1 - 2 | 44 | 62 | ||
| X57529.1 | - | 1 - 2 | 38 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_213557.1,X57529.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rps18
Genomic Map