Assay Details
| Gene Symbol: RT1-Bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004084.2 | NP_001004084.1 | 4 - 5 | 822 | 107 | |
| GenBank mRNA | BC079121.1 | - | 4 - 5 | 807 | 7126686 | 107 |
| X56596.1 | - | 4 - 5 | 761 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RT1-Bb
Genomic Map