Assay Details
| Gene Symbol: LOC103689948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256337.2 | XP_006256399.1 | 726 | 67 | ||
| Gene Symbol: Cabin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053575.2 | NP_446027.2 | 22 - 23 | 3585 | 67 | |
| GenBank mRNA | AF061947.1 | - | 22 - 23 | 3591 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103689948
Genomic Map