Assay Details
| Gene Symbol: LOC100910755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751957.3 | XP_003752005.1 | 2814 | 85 | ||
| XM_008772948.1 | XP_008771170.1 | 2233 | 85 | |||
| XM_008772949.1 | XP_008771171.1 | 2231 | 85 | |||
| Gene Symbol: Rufy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168586.1 | NP_001162058.1 | 17 - 18 | 1744 | 85 | |
| XM_006256392.2 | XP_006256454.1 | 17 - 18 | 2233 | 85 | ||
| XM_006256393.2 | XP_006256455.1 | 17 - 18 | 2231 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100910755
Genomic Map