Assay Details
| Gene Symbol: Sumo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024295.1 | NP_001019466.1 | 1 - 2 | 93 | 61 | |
| GenBank mRNA | BC083728.1 | - | 1 - 2 | 93 | 7191303 | 61 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC083728.1, NM_001024295.1 transcripts.
Set Membership: |
|

Sumo3
Genomic Map