Assay Details
| Gene Symbol: Ipmk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134417.1 | NP_599244.1 | 4 - 5 | 604 | 111 | |
| XM_006256345.2 | XP_006256407.1 | 4 - 5 | 660 | 111 | ||
| XM_006256346.2 | XP_006256408.1 | 4 - 5 | 1022 | 111 | ||
| XM_008772858.1 | XP_008771080.1 | 4 - 5 | 536 | 111 | ||
| GenBank mRNA | AY014898.1 | - | 4 - 5 | 604 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ipmk
Genomic Map