Assay Details
| Gene Symbol: RT1-Db1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008884.2 | NP_001008884.1 | 1 - 2 | 160 | 80 | |
| GenBank mRNA | AF084933.1 | - | 1 - 2 | 100 | 80 | |
| AJ003231.1 | - | 1 - 2 | 45 | 80 | ||
| AY626207.1 | - | 1 - 2 | 96 | 80 | ||
| BC087691.1 | - | 1 - 2 | 137 | 7310949 | 80 | |
| CO563535.1 | - | 1 - 2 | 160 | 80 | ||
| FQ225193.1 | - | 1 - 2 | 80 | |||
| FQ226724.1 | - | 1 - 2 | 148 | 80 | ||
| FQ226759.1 | - | 1 - 2 | 154 | 80 | ||
| FQ226850.1 | - | 1 - 2 | 148 | 80 | ||
| FQ227087.1 | - | 1 - 2 | 143 | 80 | ||
| FQ228005.1 | - | 1 - 2 | 148 | 80 | ||
| FQ228124.1 | - | 1 - 2 | 125 | 80 | ||
| FQ228129.1 | - | 1 - 2 | 148 | 80 | ||
| FQ228185.1 | - | 1 - 2 | 166 | 80 | ||
| FQ230360.1 | - | 1 - 2 | 148 | 80 | ||
| FQ231387.1 | - | 1 - 2 | 148 | 80 | ||
| FQ231709.1 | - | 1 - 2 | 148 | 80 | ||
| FQ232100.1 | - | 1 - 2 | 148 | 80 | ||
| FQ232453.1 | - | 1 - 2 | 148 | 80 | ||
| FQ233049.1 | - | 1 - 2 | 144 | 80 | ||
| FQ233335.1 | - | 1 - 2 | 148 | 80 | ||
| FQ233396.1 | - | 1 - 2 | 148 | 80 | ||
| FQ233493.1 | - | 1 - 2 | 148 | 80 | ||
| FQ233800.1 | - | 1 - 2 | 126 | 80 | ||
| FQ233801.1 | - | 1 - 2 | 148 | 80 | ||
| FQ234464.1 | - | 1 - 2 | 147 | 80 | ||
| FQ234907.1 | - | 1 - 2 | 148 | 80 | ||
| KC222950.1 | - | 1 - 2 | 96 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RT1-Db1
Genomic Map