Assay Details
| Gene Symbol: RT1-Db1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008884.2 | NP_001008884.1 | 3 - 4 | 709 | 162 | |
| GenBank mRNA | AF084933.1 | - | 3 - 4 | 649 | 162 | |
| AY626207.1 | - | 3 - 4 | 645 | 162 | ||
| BC087691.1 | - | 4 - 5 | 629 | 7310949 | 162 | |
| FQ225193.1 | - | 3 - 4 | 697 | 162 | ||
| FQ226724.1 | - | 3 - 4 | 697 | 162 | ||
| FQ226850.1 | - | 3 - 4 | 697 | 162 | ||
| FQ227087.1 | - | 3 - 4 | 694 | 162 | ||
| FQ228005.1 | - | 3 - 4 | 697 | 162 | ||
| FQ228129.1 | - | 3 - 4 | 697 | 162 | ||
| FQ228185.1 | - | 3 - 4 | 715 | 162 | ||
| FQ230360.1 | - | 3 - 4 | 697 | 162 | ||
| FQ231387.1 | - | 3 - 4 | 697 | 162 | ||
| FQ231709.1 | - | 3 - 4 | 697 | 162 | ||
| FQ232046.1 | - | 3 - 4 | 697 | 162 | ||
| FQ232100.1 | - | 3 - 4 | 696 | 162 | ||
| FQ233049.1 | - | 3 - 4 | 693 | 162 | ||
| FQ233335.1 | - | 3 - 4 | 697 | 162 | ||
| FQ234464.1 | - | 3 - 4 | 696 | 162 | ||
| KC222950.1 | - | 3 - 4 | 645 | 162 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RT1-Db1
Genomic Map