Assay Details
| Gene Symbol: Hmga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256158.2 | XP_006256220.1 | 1 - 2 | 148 | 76 | |
| XM_006256162.2 | XP_006256224.1 | 1 - 2 | 142 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006256158.2,XM_006256162.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hmga1
Genomic Map