Assay Details
| Gene Symbol: Hmga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256156.1 | XP_006256218.1 | 1 - 2 | 104 | 95 | |
| XM_006256161.1 | XP_006256223.1 | 1 - 2 | 104 | 95 | ||
| XM_006256164.1 | XP_006256226.1 | 1 - 2 | 113 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006256156.1,XM_006256161.1,XM_006256164.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hmga1
Genomic Map