Assay Details
| Gene Symbol: Lhfpl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013906.1 | NP_001013928.1 | 2 - 3 | 899 | 70 | |
| GenBank mRNA | BC087673.1 | - | 2 - 3 | 899 | 7316507 | 70 |
More Information
Updated Mapping Information
Set Membership: |
|

Lhfpl5
Genomic Map