Assay Details
| Gene Symbol: Fam13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772939.1 | XP_008771161.1 | 1 - 2 | 6907 | 56 | |
| XM_008772940.1 | XP_008771162.1 | 1 - 2 | 6907 | 56 | ||
| GenBank mRNA | FQ212194.1 | - | 671 | 56 | ||
More Information
Set Membership: |
|

Fam13c
Genomic Map