Assay Details
| Gene Symbol: Papolg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107244.1 | NP_001100714.1 | 13 - 14 | 1396 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P605593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14018 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P605593 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P605593 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P605593 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P149588 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16326 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10778791 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778783 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778778 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778795 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778784 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778793 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778774 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778782 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778779 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778787 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778777 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778790 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778775 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778789 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778788 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778786 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778792 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778780 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778794 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778776 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778781 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778785 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10778773 | - | - |
| Rat Genome 230 2.0 Array | 1397336_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380101_at | - | - |
| Rat Genome 230 2.0 Array | 1395787_at | - | - |
| Rat Genome 230 2.0 Array | 1397336_at | - | - |
| Rat Genome 230 2.0 Array | 1395787_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380101_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397475_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397475_at | - | - |
| Affymetrix HT Rat Focus Array | 1397475_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1380101_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1380101_at | - | - |
| Affymetrix HT Rat Focus Array | 1395787_at | - | - |
| Affymetrix HT Rat Focus Array | 1397336_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1397336_at | - | - |
| Affymetrix HT Rat Focus Array | 1395787_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1397475_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400017621.rn.1 | TC1400002387.rn.1 | RTA TC |
| Rat Expression Set 230 | 1395787_at | - | - |
| Rat Expression Set 230 | 1397475_PM_at | - | - |
| Rat Expression Set 230 | 1380101_at | - | - |
| Rat Expression Set 230 | 1380101_PM_at | - | - |
| Rat Expression Set 230 | 1397336_at | - | - |
| Rat Expression Set 230 | 1397336_PM_at | - | - |
| Rat Expression Set 230 | 1395787_PM_at | - | - |
| Rat Expression Set 230 | 1397475_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10778773 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20986510 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349742 | - | - |

Papolg
Genomic Map