Assay Details
| Gene Symbol: Siva1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100982.1 | NP_001094452.1 | 1 - 2 | 131 | 100 | |
| XM_006240645.2 | XP_006240707.1 | 1 - 2 | 245 | 100 | ||
| GenBank mRNA | BC088866.1 | - | 1 - 2 | 131 | 7318866 | 100 |
More Information
Updated Mapping Information
Set Membership: |
|

Siva1
Genomic Map