Assay Details
| Gene Symbol: Rd3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134560.1 | NP_001128032.1 | 1 - 2 | 387 | 125 | |
| GenBank mRNA | BC158708.1 | - | 1 - 2 | 372 | 125 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001134560.1 transcripts.
Set Membership: |
|

Rd3l
Genomic Map