Assay Details
| Gene Symbol: LOC102554884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006240612.2 | XP_006240674.1 | 8 - 9 | 878 | 65 | |
| XM_008764974.1 | XP_008763196.1 | 6 - 7 | 1767 | 65 | ||
More Information
Set Membership: |
|

LOC102554884
Genomic Map