Assay Details
| Gene Symbol: Cep170b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729727.4 | XP_002729773.2 | 3 - 4 | 1307 | 65 | |
| XM_008764990.1 | XP_008763212.1 | 3 - 4 | 2120 | 65 | ||
| XM_008764991.1 | XP_008763213.1 | 4 - 5 | 1959 | 65 | ||
| XM_576105.5 | XP_576105.4 | 3 - 4 | 1307 | 65 | ||
More Information
Set Membership: |
|

Cep170b
Genomic Map