Assay Details
| Gene Symbol: LOC100910833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008764908.1 | XP_008763130.1 | 4 - 5 | 2295 | 84 | |
| Gene Symbol: Setd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_216781.8 | XP_216781.6 | 7 - 8 | 892 | 84 | |
| GenBank mRNA | BC089108.1 | - | 2 - 3 | 292 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100910833
Genomic Map