Assay Details
| Gene Symbol: Cdca4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037214.1 | NP_001032291.1 | 1 - 2 | 277 | 56 | |
| GenBank mRNA | BC107668.1 | - | 1 - 2 | 277 | 7937865 | 56 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC107668.1, NM_001037214.1 transcripts.
Set Membership: |
|

Cdca4
Genomic Map