Assay Details
| Gene Symbol: Ipcef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170799.1 | NP_001164270.1 | 1 - 2 | 136 | 111 | |
| GenBank mRNA | AJ536192.1 | - | 1 - 2 | 136 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ536192.1, NM_001170799.1 transcripts.
Set Membership: |
|

Ipcef1
Genomic Map