Assay Details
| Gene Symbol: Ipcef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170799.1 | NP_001164270.1 | 3 - 4 | 374 | 83 | |
| GenBank mRNA | AJ536192.1 | - | 3 - 4 | 374 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ipcef1
Genomic Map