Assay Details
| Gene Symbol: Timp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053819.1 | NP_446271.1 | 4 - 5 | 363 | 61 | |
| XM_006256608.2 | XP_006256670.1 | 4 - 5 | 427 | 61 | ||
| XM_008773034.1 | XP_008771256.1 | 3 - 4 | 337 | 61 | ||
| GenBank mRNA | AF411319.1 | - | 1 - 2 | 113 | 61 | |
| AY550026.1 | - | 3 - 4 | 352 | 61 | ||
| BC099821.1 | - | 4 - 5 | 361 | 7930800 | 61 | |
| FQ219856.1 | - | 4 - 5 | 373 | 61 | ||
| FQ222271.1 | - | 4 - 5 | 373 | 61 | ||
| FQ222437.1 | - | 4 - 5 | 376 | 61 | ||
| FQ222915.1 | - | 4 - 5 | 373 | 61 | ||
| FQ228400.1 | - | 4 - 5 | 372 | 61 | ||
| FQ230039.1 | - | 4 - 5 | 370 | 61 | ||
| L31883.1 | - | 4 - 5 | 363 | 61 | ||
| U06179.1 | - | 3 - 4 | 346 | 61 | ||
| U16022.1 | - | 2 - 3 | 257 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Timp1
Genomic Map