Assay Details
| Gene Symbol: Mon1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255674.2 | XP_006255736.1 | 2 - 3 | 524 | 62 | |
| XM_006255675.2 | XP_006255737.1 | 2 - 3 | 338 | 62 | ||
| XM_008772515.1 | XP_008770737.1 | 1 - 2 | 553 | 62 | ||
More Information
Set Membership: |
|

Mon1b
Genomic Map