Assay Details
| Gene Symbol: Dpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008383.1 | NP_001008384.1 | 1 - 2 | 391 | 77 | |
| XM_006255517.2 | XP_006255579.1 | 2 - 3 | 412 | 77 | ||
| XM_008772521.1 | XP_008770743.1 | 2 - 3 | 609 | 77 | ||
| GenBank mRNA | BC085826.1 | - | 1 - 2 | 391 | 7131855 | 77 |
More Information
Updated Mapping Information
Set Membership: |
|

Dpep3
Genomic Map