Assay Details
| Gene Symbol: Dpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008383.1 | NP_001008384.1 | 2 - 3 | 523 | 128 | |
| XM_006255517.2 | XP_006255579.1 | 3 - 4 | 544 | 128 | ||
| XM_008772521.1 | XP_008770743.1 | 3 - 4 | 741 | 128 | ||
| GenBank mRNA | BC085826.1 | - | 2 - 3 | 523 | 7131855 | 128 |
More Information
Updated Mapping Information
Set Membership: |
|

Dpep3
Genomic Map