Assay Details
| Gene Symbol: Tcf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751893.3 | XP_003751941.1 | 12 - 13 | 1691 | 61 | |
| XM_006255791.2 | XP_006255853.1 | 12 - 13 | 1691 | 61 | ||
| XM_006255792.2 | XP_006255854.1 | 12 - 13 | 1691 | 61 | ||
| XM_006255793.2 | XP_006255855.1 | 11 - 12 | 1619 | 61 | ||
| XM_006255794.1 | XP_006255856.1 | 12 - 13 | 1691 | 61 | ||
| XM_214707.8 | XP_214707.6 | 12 - 13 | 1691 | 61 | ||
| GenBank mRNA | FQ215148.1 | - | 12 - 13 | 1457 | 61 | |
| FQ230504.1 | - | 12 - 13 | 1437 | 61 | ||
More Information
Set Membership: |
|

Tcf25
Genomic Map