Assay Details
| Gene Symbol: Gab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108444.1 | NP_001101914.1 | 8 - 9 | 2082 | 63 | |
| GenBank mRNA | GQ140625.1 | - | 9 - 10 | 1882 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
response to oxidative stress
signal transduction
epidermal growth factor receptor signaling pathway
activation of JUN kinase activity
heart development
epidermis development
regulation of cell migration
positive regulation of MAPK cascade
platelet-derived growth factor receptor signaling pathway
labyrinthine layer development
interleukin-6-mediated signaling pathway
signal transducer activity
activation of MAPK activity
response to oxidative stress
signal transduction
epidermal growth factor receptor signaling pathway
activation of JUN kinase activity
heart development
epidermis development
regulation of cell migration
positive regulation of MAPK cascade
platelet-derived growth factor receptor signaling pathway
labyrinthine layer development
interleukin-6-mediated signaling pathway
signal transducer activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P494608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11153 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P494608 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P494608 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P494608 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P494608 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52593 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90120 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53380 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10806936 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10806930 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10806933 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10806928 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10806932 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10806935 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10806934 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10806929 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10806931 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10806927 | - | - |
| Rat Genome 230 2.0 Array | 1399167_a_at | - | - |
| Rat Genome 230 2.0 Array | 1399167_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1399167_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1399167_a_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1900003238.rn.1 | TC1900000408.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900003236.rn.1 | TC1900000408.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900003237.rn.1 | TC1900000408.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10806927 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20889210 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349894 | - | - |

Gab1
Genomic Map