Assay Details
| Gene Symbol: RGD1304884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107436.1 | NP_001100906.1 | 2 - 3 | 609 | 93 | |
| XM_006255743.2 | XP_006255805.1 | 1 - 2 | 457 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RGD1304884
Genomic Map