Assay Details
| Gene Symbol: Map1lc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022867.2 | NP_074058.2 | 2 - 3 | 170 | 82 | |
| GenBank mRNA | AY206669.1 | - | 2 - 3 | 132 | 82 | |
| AY392036.1 | - | 2 - 3 | 170 | 82 | ||
| BC058144.1 | - | 2 - 3 | 128 | 6889616 | 82 | |
| BC083556.1 | - | 2 - 3 | 150 | 7189855 | 82 | |
| FQ221245.1 | - | 2 - 3 | 181 | 82 | ||
| FQ228912.1 | - | 2 - 3 | 181 | 82 | ||
| U05784.1 | - | 2 - 3 | 120 | 82 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Map1lc3b
Genomic Map